Learn More
Hampton Research PEGRx 2, 10ml
SDP
Catalog No. 500933
Encompass
Crystallization reagent kit designed to evaluate an array of polymers of varying molecular weight in a medium ionic strength environment in the presence of additives, salts, volatile organics and polyols versus a wide range of pH. Polymer reagents include Polyethylene glycols and Polyethylene glycol monomethyl ethers. The polymer molecular weight range between 200 and 20,000 is evaluated in a medium ionic strength formulation. Ten different buffers are used to span the range of pH between 3.5 and 9. The primary screen variables are polymer type, polymer molecular weight, pH and secondary reagents which include additives, salts, volatile organics and polyols. 10mlPlease note items from this supplier are non-returnable.
Catalog No. 50-093-3 Supplier Hampton Research Supplier No. HR2084
Crystallization reagent kit designed to evaluate an array of polymers of varying molecular weight in a medium ionic strength environment in the presence of additives, salts, volatile organics and polyols versus a wide range of pH. Polymer reagents include Polyethylene glycols and Polyethylene glycol monomethyl ethers. The polymer molecular weight range between 200 and 20,000 is evaluated in a medium ionic strength formulation. Ten different buffers are used to span the range of pH between 3.5 and 9. The primary screen variables are polymer type, polymer molecular weight, pH and secondary reagents which include additives, salts, volatile organics and polyols. 10mlPlease note items from this supplier are non-returnable.