Learn More
Hampton Research HR2-984-01/PEGRx 2 #01 (E01), 185 ml - 0.8 M Lithium sulfate monohydrate, 0.1 M Sodium acetate trihydrate pH 4.0, 4% v/v Polyethylene glycol 200
SDP
Catalog No. 502342
Encompass
HR2-984-01/PEGRx 2 #01 (E01), 185 ml - 0.8 M Lithium sulfate monohydrate, 0.1 M Sodium acetate trihydrate pH 4.0, 4% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.
Catalog No. 50-234-2 Supplier Hampton Research Supplier No. HR298401
HR2-984-01/PEGRx 2 #01 (E01), 185 ml - 0.8 M Lithium sulfate monohydrate, 0.1 M Sodium acetate trihydrate pH 4.0, 4% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.