Learn More
Hampton Research HR2-984-02/PEGRx 2 #02 (E02), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 26% v/v Polyethylene glycol 200
SDP
Supplier:  Hampton Research HR298402
Encompass
HR2-984-02/PEGRx 2 #02 (E02), 185 ml - 0.2 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 26% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.
Catalog No. 50-234-3