Learn More
Hampton Research HR2-984-03/PEGRx 2 #03 (E03), 185 ml - 0.05 M Calcium chloride dihydrate, 0.1 M MES monohydrate pH 6.0, 45% v/v Polyethylene glycol 200
SDP
Catalog No. 502344
Encompass
HR2-984-03/PEGRx 2 #03 (E03), 185 ml - 0.05 M Calcium chloride dihydrate, 0.1 M MES monohydrate pH 6.0, 45% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.
Catalog No. 50-234-4 Supplier Hampton Research Supplier No. HR298403
HR2-984-03/PEGRx 2 #03 (E03), 185 ml - 0.05 M Calcium chloride dihydrate, 0.1 M MES monohydrate pH 6.0, 45% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.