Learn More
Hampton Research HR2-984-04/PEGRx 2 #04 (E04), 185 ml - 28% v/v 2-Propanol, 0.1 M BIS-TRIS pH 6.5, 3% v/v Polyethylene glycol 200
SDP
Catalog No. 502345
Encompass
HR2-984-04/PEGRx 2 #04 (E04), 185 ml - 28% v/v 2-Propanol, 0.1 M BIS-TRIS pH 6.5, 3% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.
Catalog No. 50-234-5 Supplier Hampton Research Supplier No. HR298404
HR2-984-04/PEGRx 2 #04 (E04), 185 ml - 28% v/v 2-Propanol, 0.1 M BIS-TRIS pH 6.5, 3% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.