Learn More
Hampton Research HR2-984-05/PEGRx 2 #05 (E05), 185 ml - 20% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.5, 2% v/v Polyethylene glycol 200
SDP
Catalog No. 502346
Encompass
HR2-984-05/PEGRx 2 #05 (E05), 185 ml - 20% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.5, 2% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.
Catalog No. 50-234-6 Supplier Hampton Research Supplier No. HR298405
Add to Cart
Add to Cart
HR2-984-05/PEGRx 2 #05 (E05), 185 ml - 20% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.5, 2% v/v Polyethylene glycol 200. Please note items from this supplier are non-returnable.