Learn More
Hampton Research HR2-984-07/PEGRx 2 #07 (E07), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 24% v/v Polyethylene glycol 400
SDP
Catalog No. 502348
Encompass
HR2-984-07/PEGRx 2 #07 (E07), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 24% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-234-8 Supplier Hampton Research Supplier No. HR298407
HR2-984-07/PEGRx 2 #07 (E07), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 24% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.