Learn More
Hampton Research HR2-984-08/PEGRx 2 #08 (E08), 185 ml - 0.2 M Ammonium sulfate, 0.1 M BIS-TRIS pH 6.5, 18% v/v Polyethylene glycol 400
SDP
Catalog No. 502349
Encompass
HR2-984-08/PEGRx 2 #08 (E08), 185 ml - 0.2 M Ammonium sulfate, 0.1 M BIS-TRIS pH 6.5, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-234-9 Supplier Hampton Research Supplier No. HR298408
HR2-984-08/PEGRx 2 #08 (E08), 185 ml - 0.2 M Ammonium sulfate, 0.1 M BIS-TRIS pH 6.5, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.