Learn More
Hampton Research HR2-984-09/PEGRx 2 #09 (E09), 185 ml - 0.19 mM CYMAL-7, 0.1 M HEPES pH 7.5, 40% v/v Polyethylene glycol 400
SDP
Catalog No. 502350
Encompass
HR2-984-09/PEGRx 2 #09 (E09), 185 ml - 0.19 mM CYMAL-7, 0.1 M HEPES pH 7.5, 40% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-235-0 Supplier Hampton Research Supplier No. HR298409
HR2-984-09/PEGRx 2 #09 (E09), 185 ml - 0.19 mM CYMAL-7, 0.1 M HEPES pH 7.5, 40% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.