Learn More
Hampton Research HR2-984-10/PEGRx 2 #10 (E10), 185 ml - 6% v/v 2-Propanol, 0.1 M Sodium acetate trihydrate pH 4.5, 26% v/v Polyethylene glycol monomethyl ether 550
SDP
Catalog No. 502351
Encompass
HR2-984-10/PEGRx 2 #10 (E10), 185 ml - 6% v/v 2-Propanol, 0.1 M Sodium acetate trihydrate pH 4.5, 26% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.
Catalog No. 50-235-1 Supplier Hampton Research Supplier No. HR298410
HR2-984-10/PEGRx 2 #10 (E10), 185 ml - 6% v/v 2-Propanol, 0.1 M Sodium acetate trihydrate pH 4.5, 26% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.