Learn More
Hampton Research HR2-984-12/PEGRx 2 #12 (E12), 185 ml - 0.15 M DL-Malic acid pH 7.0, 0.1 M Imidazole pH 7.0, 22% v/v Polyethylene glycol monomethyl ether 550
SDP
Catalog No. 502353
Encompass
HR2-984-12/PEGRx 2 #12 (E12), 185 ml - 0.15 M DL-Malic acid pH 7.0, 0.1 M Imidazole pH 7.0, 22% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.
Catalog No. 50-235-3 Supplier Hampton Research Supplier No. HR298412
HR2-984-12/PEGRx 2 #12 (E12), 185 ml - 0.15 M DL-Malic acid pH 7.0, 0.1 M Imidazole pH 7.0, 22% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.