Learn More
Hampton Research HR2-984-13/PEGRx 2 #13 (F01), 185 ml - 0.1 M Succinic acid pH 7.0, 0.1 M BICINE pH 8.5, 30% v/v Polyethylene glycol monomethyl ether 550
SDP
Catalog No. 502354
Encompass
HR2-984-13/PEGRx 2 #13 (F01), 185 ml - 0.1 M Succinic acid pH 7.0, 0.1 M BICINE pH 8.5, 30% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.
Catalog No. 50-235-4 Supplier Hampton Research Supplier No. HR298413
HR2-984-13/PEGRx 2 #13 (F01), 185 ml - 0.1 M Succinic acid pH 7.0, 0.1 M BICINE pH 8.5, 30% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.