Learn More
Hampton Research HR2-984-17/PEGRx 2 #17 (F05), 185 ml - 0.2 M L-Proline, 0.1 M HEPES pH 7.5, 24% w/v Polyethylene glycol 1,500
SDP
Catalog No. 502358
Encompass
HR2-984-17/PEGRx 2 #17 (F05), 185 ml - 0.2 M L-Proline, 0.1 M HEPES pH 7.5, 24% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.
Catalog No. 50-235-8 Supplier Hampton Research Supplier No. HR298417
HR2-984-17/PEGRx 2 #17 (F05), 185 ml - 0.2 M L-Proline, 0.1 M HEPES pH 7.5, 24% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.