Learn More
Hampton Research HR2-984-18/PEGRx 2 #18 (F06), 185 ml - 10% v/v 2-Propanol, 0.1 M BICINE pH 8.5, 30% w/v Polyethylene glycol 1,500
SDP
Catalog No. 502359
Encompass
HR2-984-18/PEGRx 2 #18 (F06), 185 ml - 10% v/v 2-Propanol, 0.1 M BICINE pH 8.5, 30% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.
Catalog No. 50-235-9 Supplier Hampton Research Supplier No. HR298418
HR2-984-18/PEGRx 2 #18 (F06), 185 ml - 10% v/v 2-Propanol, 0.1 M BICINE pH 8.5, 30% w/v Polyethylene glycol 1,500. Please note items from this supplier are non-returnable.