Learn More
Hampton Research HR2-984-20/PEGRx 2 #20 (F08), 185 ml - 0.02 M Nickel(II) chloride hexahydrate, 0.02 M Magnesium chloride hexahydrate, 0.02 M Cadmium chloride hydrate, 0.1 M Sodium acetate trihydrate pH 4.5, 24% w/v PEG monomethyl ether 2,000
SDP
Catalog No. 502361
Encompass
HR2-984-20/PEGRx 2 #20 (F08), 185 ml - 0.02 M Nickel(II) chloride hexahydrate, 0.02 M Magnesium chloride hexahydrate, 0.02 M Cadmium chloride hydrate, 0.1 M Sodium acetate trihydrate pH 4.5, 24% w/v PEG monomethyl ether 2,000. Please note items from this supplier are non-returnable.
Catalog No. 50-236-1 Supplier Hampton Research Supplier No. HR298420
HR2-984-20/PEGRx 2 #20 (F08), 185 ml - 0.02 M Nickel(II) chloride hexahydrate, 0.02 M Magnesium chloride hexahydrate, 0.02 M Cadmium chloride hydrate, 0.1 M Sodium acetate trihydrate pH 4.5, 24% w/v PEG monomethyl ether 2,000. Please note items from this supplier are non-returnable.