Learn More
Hampton Research HR2-984-21/PEGRx 2 #21 (F09), 185 ml - 20% v/v 2-Propanol, 0.1 M MES monohydrate pH 6.0, 20% w/v Polyethylene glycol monomethyl ether 2,000
SDP
Catalog No. 502362
Encompass
HR2-984-21/PEGRx 2 #21 (F09), 185 ml - 20% v/v 2-Propanol, 0.1 M MES monohydrate pH 6.0, 20% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.
Catalog No. 50-236-2 Supplier Hampton Research Supplier No. HR298421
HR2-984-21/PEGRx 2 #21 (F09), 185 ml - 20% v/v 2-Propanol, 0.1 M MES monohydrate pH 6.0, 20% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.