Learn More
Hampton Research HR2-984-24/PEGRx 2 #24 (F12), 185 ml - 50% v/v Tacsimate pH 4.0, 0.1 M Sodium acetate trihydrate pH 4.5, 1% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502365
Encompass
HR2-984-24/PEGRx 2 #24 (F12), 185 ml - 50% v/v Tacsimate pH 4.0, 0.1 M Sodium acetate trihydrate pH 4.5, 1% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-236-5 Supplier Hampton Research Supplier No. HR298424
HR2-984-24/PEGRx 2 #24 (F12), 185 ml - 50% v/v Tacsimate pH 4.0, 0.1 M Sodium acetate trihydrate pH 4.5, 1% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.