Learn More
Hampton Research HR2-984-25/PEGRx 2 #25 (G01), 185 ml - 0.10% w/v n-Octyl-B-D-glucoside, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 22% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502366
Encompass
HR2-984-25/PEGRx 2 #25 (G01), 185 ml - 0.10% w/v n-Octyl-B-D-glucoside, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 22% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-236-6 Supplier Hampton Research Supplier No. HR298425
HR2-984-25/PEGRx 2 #25 (G01), 185 ml - 0.10% w/v n-Octyl-B-D-glucoside, 0.1 M Sodium citrate tribasic dihydrate pH 5.5, 22% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.