Learn More
Hampton Research HR2-984-26/PEGRx 2 #26 (G02), 185 ml - 2% v/v Tacsimate pH 7.0, 5% v/v 2-Propanol, 0.1 M Imidazole pH 7.0, 8% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502367
Encompass
HR2-984-26/PEGRx 2 #26 (G02), 185 ml - 2% v/v Tacsimate pH 7.0, 5% v/v 2-Propanol, 0.1 M Imidazole pH 7.0, 8% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-236-7 Supplier Hampton Research Supplier No. HR298426
HR2-984-26/PEGRx 2 #26 (G02), 185 ml - 2% v/v Tacsimate pH 7.0, 5% v/v 2-Propanol, 0.1 M Imidazole pH 7.0, 8% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.