Learn More
Hampton Research HR2-984-29/PEGRx 2 #29 (G05), 185 ml - 6% v/v Tacsimate pH 6.0, 0.1 M MES monohydrate pH 6.0, 25% w/v Polyethylene glycol 4,000
SDP
Catalog No. 502370
Encompass
HR2-984-29/PEGRx 2 #29 (G05), 185 ml - 6% v/v Tacsimate pH 6.0, 0.1 M MES monohydrate pH 6.0, 25% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-0 Supplier Hampton Research Supplier No. HR298429
HR2-984-29/PEGRx 2 #29 (G05), 185 ml - 6% v/v Tacsimate pH 6.0, 0.1 M MES monohydrate pH 6.0, 25% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.