Learn More
Hampton Research HR2-984-30/PEGRx 2 #30 (G06), 185 ml - 0.2 M Magnesium formate dihydrate, 0.1 M Sodium acetate trihydrate pH 4.0, 18% w/v Polyethylene glycol monomethyl ether 5,000
SDP
Catalog No. 502371
Encompass
HR2-984-30/PEGRx 2 #30 (G06), 185 ml - 0.2 M Magnesium formate dihydrate, 0.1 M Sodium acetate trihydrate pH 4.0, 18% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-1 Supplier Hampton Research Supplier No. HR298430
HR2-984-30/PEGRx 2 #30 (G06), 185 ml - 0.2 M Magnesium formate dihydrate, 0.1 M Sodium acetate trihydrate pH 4.0, 18% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.