Learn More
Hampton Research HR2-984-31/PEGRx 2 #31 (G07), 185 ml - 2% v/v Polyethylene glycol 400, 0.1 M Imidazole pH 7.0, 24% w/v Polyethylene glycol monomethyl ether 5,000
SDP
Catalog No. 502372
Encompass
HR2-984-31/PEGRx 2 #31 (G07), 185 ml - 2% v/v Polyethylene glycol 400, 0.1 M Imidazole pH 7.0, 24% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-2 Supplier Hampton Research Supplier No. HR298431
HR2-984-31/PEGRx 2 #31 (G07), 185 ml - 2% v/v Polyethylene glycol 400, 0.1 M Imidazole pH 7.0, 24% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.