Learn More
Hampton Research HR2-984-32/PEGRx 2 #32 (G08), 185 ml - 0.2 M Sodium formate, 0.1 M BICINE pH 8.5, 20% w/v Polyethylene glycol monomethyl ether 5,000
SDP
Catalog No. 502373
Encompass
HR2-984-32/PEGRx 2 #32 (G08), 185 ml - 0.2 M Sodium formate, 0.1 M BICINE pH 8.5, 20% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-3 Supplier Hampton Research Supplier No. HR298432
HR2-984-32/PEGRx 2 #32 (G08), 185 ml - 0.2 M Sodium formate, 0.1 M BICINE pH 8.5, 20% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.