Learn More
Hampton Research HR2-984-33/PEGRx 2 #33 (G09), 185 ml - 4% v/v 2-Propanol, 0.1 M BIS-TRIS propane pH 9.0, 20% w/v Polyethylene glycol monomethyl ether 5,000
SDP
Catalog No. 502374
Encompass
HR2-984-33/PEGRx 2 #33 (G09), 185 ml - 4% v/v 2-Propanol, 0.1 M BIS-TRIS propane pH 9.0, 20% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-4 Supplier Hampton Research Supplier No. HR298433
Add to Cart
Add to Cart
HR2-984-33/PEGRx 2 #33 (G09), 185 ml - 4% v/v 2-Propanol, 0.1 M BIS-TRIS propane pH 9.0, 20% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.