Learn More
Hampton Research HR2-984-34/PEGRx 2 #34 (G10), 185 ml - 6% v/v Ethylene glycol, 0.1 M Citric acid pH 3.5, 10% w/v Polyethylene glycol 6,000
SDP
Catalog No. 502375
Encompass
HR2-984-34/PEGRx 2 #34 (G10), 185 ml - 6% v/v Ethylene glycol, 0.1 M Citric acid pH 3.5, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-5 Supplier Hampton Research Supplier No. HR298434
HR2-984-34/PEGRx 2 #34 (G10), 185 ml - 6% v/v Ethylene glycol, 0.1 M Citric acid pH 3.5, 10% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.