Learn More
Hampton Research HR2-984-35/PEGRx 2 #35 (G11), 185 ml - 0.15 M Lithium sulfate monohydrate, 0.1 M Citric acid pH 3.5, 18% Polyethylene glycol 6,000
SDP
Catalog No. 502376
Encompass
HR2-984-35/PEGRx 2 #35 (G11), 185 ml - 0.15 M Lithium sulfate monohydrate, 0.1 M Citric acid pH 3.5, 18% Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-6 Supplier Hampton Research Supplier No. HR298435
HR2-984-35/PEGRx 2 #35 (G11), 185 ml - 0.15 M Lithium sulfate monohydrate, 0.1 M Citric acid pH 3.5, 18% Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.