Learn More
Hampton Research HR2-984-36/PEGRx 2 #36 (G12), 185 ml - 10% v/v 2-Propanol, 0.1 M Sodium acetate trihydrate pH 4.0, 22% w/v Polyethylene glycol 6,000
SDP
Catalog No. 502377
Encompass
HR2-984-36/PEGRx 2 #36 (G12), 185 ml - 10% v/v 2-Propanol, 0.1 M Sodium acetate trihydrate pH 4.0, 22% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-7 Supplier Hampton Research Supplier No. HR298436
HR2-984-36/PEGRx 2 #36 (G12), 185 ml - 10% v/v 2-Propanol, 0.1 M Sodium acetate trihydrate pH 4.0, 22% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.