Learn More
Hampton Research HR2-984-37/PEGRx 2 #37 (H01), 185 ml - 0.2 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.0, 22% w/v Polyethylene glycol 8,000
SDP
Catalog No. 502378
Encompass
HR2-984-37/PEGRx 2 #37 (H01), 185 ml - 0.2 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.0, 22% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-8 Supplier Hampton Research Supplier No. HR298437
HR2-984-37/PEGRx 2 #37 (H01), 185 ml - 0.2 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.0, 22% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.