Learn More
Hampton Research HR2-984-38/PEGRx 2 #38 (H02), 185 ml - 20% v/v 2-Propanol, 0.1 M Tris pH 8.0, 5% w/v Polyethylene glycol 8,000
SDP
Catalog No. 502379
Encompass
HR2-984-38/PEGRx 2 #38 (H02), 185 ml - 20% v/v 2-Propanol, 0.1 M Tris pH 8.0, 5% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-237-9 Supplier Hampton Research Supplier No. HR298438
HR2-984-38/PEGRx 2 #38 (H02), 185 ml - 20% v/v 2-Propanol, 0.1 M Tris pH 8.0, 5% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.