Learn More
Hampton Research HR2-984-39/PEGRx 2 #39 (H03), 185 ml - 10% v/v Polyethylene glycol 200, 0.1 M BIS-TRIS propane pH 9.0, 18% w/v Polyethylene glycol 8,000
SDP
Catalog No. 502380
Encompass
HR2-984-39/PEGRx 2 #39 (H03), 185 ml - 10% v/v Polyethylene glycol 200, 0.1 M BIS-TRIS propane pH 9.0, 18% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-238-0 Supplier Hampton Research Supplier No. HR298439
HR2-984-39/PEGRx 2 #39 (H03), 185 ml - 10% v/v Polyethylene glycol 200, 0.1 M BIS-TRIS propane pH 9.0, 18% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.