Learn More
Hampton Research HR2-984-40/PEGRx 2 #40 (H04), 185 ml - 15% v/v 2-Propanol, 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 10% w/v Polyethylene glycol 10,000
SDP
Catalog No. 502381
Encompass
HR2-984-40/PEGRx 2 #40 (H04), 185 ml - 15% v/v 2-Propanol, 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 10% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.
Catalog No. 50-238-1 Supplier Hampton Research Supplier No. HR298440
HR2-984-40/PEGRx 2 #40 (H04), 185 ml - 15% v/v 2-Propanol, 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 10% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.