Learn More
Hampton Research HR2-984-41/PEGRx 2 #41 (H05), 185 ml - 0.4 M Sodium malonate pH 6.0, 0.1 M MES monohydrate pH 6.0, 0.5% w/v Polyethylene glycol 10,000
SDP
Catalog No. 502382
Encompass
HR2-984-41/PEGRx 2 #41 (H05), 185 ml - 0.4 M Sodium malonate pH 6.0, 0.1 M MES monohydrate pH 6.0, 0.5% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.
Catalog No. 50-238-2 Supplier Hampton Research Supplier No. HR298441
HR2-984-41/PEGRx 2 #41 (H05), 185 ml - 0.4 M Sodium malonate pH 6.0, 0.1 M MES monohydrate pH 6.0, 0.5% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.