Learn More
Hampton Research HR2-984-44/PEGRx 2 #44 (H08), 185 ml - 0.2 M Ammonium acetate, 0.1 M Tris pH 8.0, 16% w/v Polyethylene glycol 10,000
SDP
Catalog No. 502385
Encompass
HR2-984-44/PEGRx 2 #44 (H08), 185 ml - 0.2 M Ammonium acetate, 0.1 M Tris pH 8.0, 16% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.
Catalog No. 50-238-5 Supplier Hampton Research Supplier No. HR298444
HR2-984-44/PEGRx 2 #44 (H08), 185 ml - 0.2 M Ammonium acetate, 0.1 M Tris pH 8.0, 16% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.