Learn More
Hampton Research HR2-984-45/PEGRx 2 #45 (H09), 185 ml - 5% v/v 2-Propanol, 0.1 M Citric acid pH 3.5, 6% w/v Polyethylene glycol 20,000
SDP
Supplier:  Hampton Research HR298445
Encompass
HR2-984-45/PEGRx 2 #45 (H09), 185 ml - 5% v/v 2-Propanol, 0.1 M Citric acid pH 3.5, 6% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.
Catalog No. 50-238-6