Learn More
Hampton Research HR2-984-46/PEGRx 2 #46 (H10), 185 ml - 1.0 M Sodium malonate pH 5.0, 0.1 M Sodium acetate trihydrate pH 4.5, 2% w/v Polyethylene glycol 20,000
SDP
Catalog No. 502387
Encompass
HR2-984-46/PEGRx 2 #46 (H10), 185 ml - 1.0 M Sodium malonate pH 5.0, 0.1 M Sodium acetate trihydrate pH 4.5, 2% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.
Catalog No. 50-238-7 Supplier Hampton Research Supplier No. HR298446
HR2-984-46/PEGRx 2 #46 (H10), 185 ml - 1.0 M Sodium malonate pH 5.0, 0.1 M Sodium acetate trihydrate pH 4.5, 2% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.