Learn More
Hampton Research HR2-984-47/PEGRx 2 #47 (H11), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 10% w/v Polyethylene glycol 20,000
SDP
Catalog No. 502388
Encompass
HR2-984-47/PEGRx 2 #47 (H11), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 10% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.
Catalog No. 50-238-8 Supplier Hampton Research Supplier No. HR298447
HR2-984-47/PEGRx 2 #47 (H11), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.0, 10% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.