Learn More
Medchemexpress LLC Pegsebrenatide | 2243292-26-2 | 95.5% | 1 MG
SDP
Catalog No. 5000425004
Encompass_Preferred
Pegsebrenatide (NLY01) is a blood-brain barrier-penetrant GLP-1R agonist. It alleviates retinal inflammation and neuronal death, delays onset and reduces disease severity in experimental autoimmune encephalomyelitis, and exerts neuroprotective effects in Parkinson's disease models by preventing microglia-mediated conversion of astrocytes to the A1 neurotoxic phenotype. It is useful for research on glaucoma, Parkinson's disease, and multiple sclerosis.
  • Blood-brain barrier-penetrant GLP-1R agonist
  • Alleviates retinal inflammation and neuron death
  • Delays onset and reduces disease severity in experimental autoimmune encephalomyelitis
  • Inhibits formation of A1 reactive astrocytes
  • Reduces loss of retinal ganglion cells and dopaminergic neurons
  • Exerts neuroprotective effects in Parkinson's disease models
  • Useful for research on glaucoma, Parkinson's disease, and multiple sclerosis
Catalog No. 50-004-25004 Supplier Medchemexpress LLC Supplier No. HYP100191MG
Pegsebrenatide (NLY01) is a blood-brain barrier-penetrant GLP-1R agonist. It alleviates retinal inflammation and neuronal death, delays onset and reduces disease severity in experimental autoimmune encephalomyelitis, and exerts neuroprotective effects in Parkinson's disease models by preventing microglia-mediated conversion of astrocytes to the A1 neurotoxic phenotype. It is useful for research on glaucoma, Parkinson's disease, and multiple sclerosis.
  • Blood-brain barrier-penetrant GLP-1R agonist
  • Alleviates retinal inflammation and neuron death
  • Delays onset and reduces disease severity in experimental autoimmune encephalomyelitis
  • Inhibits formation of A1 reactive astrocytes
  • Reduces loss of retinal ganglion cells and dopaminergic neurons
  • Exerts neuroprotective effects in Parkinson's disease models
  • Useful for research on glaucoma, Parkinson's disease, and multiple sclerosis