Learn More
Creative Biomart Recombinant HCV Antigen
Supplier:  Creative Biomart THP0331
Encompass_Preferred
Recombinant HCV protein produced in E. coli containing the Core (130 a.a.) and NS3 (236 a.a.) excluding NS4 and NS5 domains having a total Mw of 45 kDa and purified by proprietary chromatographic tech
Catalog No. 50-208-4844
 Restricted Item