Learn More
Creative Biomart Pegaspargase
Supplier:  Creative Biomart THP0087
Encompass_Preferred
Pegylated L-asparagine amidohydrolase from E. coli. Pegylation substantially (by a factor of 4) extends the protein half life.
Catalog No. 50-208-4138