Learn More
CTL Scientific Supply Corp PEHANON INDICATOR STRIPS 5.2-6
SDP
Supplier:  CTL Scientific Supply Corp 90416
Encompass_Preferred
Pehannon Indicator Strips where the indicator and color scale are combined on a single strip. Gradation of scale: 5.2 - 5.5 - 5.7 - 5.9 - 6.1 - 6.3 - 6.5 - 6.8 200 strips/box
Catalog No. 50-300-342