Learn More
CTL Scientific Supply Corp PEHANON INDICATOR STRIPS 8.0-9
SDP
Supplier:  CTL Scientific Supply Corp 90420
Encompass_Preferred
Pehannon Indicator Strips where the indicator and color scale are combined on a single strip. Gradation of scale: 8.0 - 8.2 - 8.4 - 8.6 - 8.8 - 9.1 - 9.4 - 9.7 200/strips box
Catalog No. 50-300-343