Learn More
Polysciences, Inc. Polyethylenimine, Branched, Mw 70,000, 30% w/v aq. soln. (bPEI 70000 30% soln.)
SDP
Catalog No. NC1929722
Encompass_Preferred
Branched Polyethylenimine,, Mw 70,000, 30% w/v aq. soln. (bPEI 70000 soln.) is highly branched polyamine with high cationic charge density. bPEI 70000 contains primary, secondary, and tertiary amine groups in approximately 25/50/25 ratio.Industrially, branched polyethylenimine (bPEI) have extensive applications as an auxiliary agent intensifying manufacturing processes and improving quality of the final products. They are widely used in: paper-making, water treatment, detergents, adhesives, and cosmetics. In research, they are extensively investigated as non-viral vector carriers.
Catalog No. NC1929722 Supplier Polysciences, Inc. Supplier No. 06090500
Add to Cart
Add to Cart
Branched Polyethylenimine,, Mw 70,000, 30% w/v aq. soln. (bPEI 70000 soln.) is highly branched polyamine with high cationic charge density. bPEI 70000 contains primary, secondary, and tertiary amine groups in approximately 25/50/25 ratio.Industrially, branched polyethylenimine (bPEI) have extensive applications as an auxiliary agent intensifying manufacturing processes and improving quality of the final products. They are widely used in: paper-making, water treatment, detergents, adhesives, and cosmetics. In research, they are extensively investigated as non-viral vector carriers.