Learn More
HANNA INSTRUMENTS INC Refillable PEI Body pH Electrode with BNC Connector
SDP
Catalog No. NC1846013
Encompass
The HI1332B is a PEI body, double junction pH electrode with a BNC connector. This electrode has a single ceramic junction in the outer reference cell and the pH sensing portion is made with general purpose glass, protected by an extension of the durable PEI body. This design consideration is ideal for chemicals, quality control, field applications, and general purpose use.Durable PEI BodyDouble Junction DesignRefillable Electrode
Catalog No. NC1846013 Supplier HANNA INSTRUMENTS INC Supplier No. HI1332B
The HI1332B is a PEI body, double junction pH electrode with a BNC connector. This electrode has a single ceramic junction in the outer reference cell and the pH sensing portion is made with general purpose glass, protected by an extension of the durable PEI body. This design consideration is ideal for chemicals, quality control, field applications, and general purpose use.Durable PEI BodyDouble Junction DesignRefillable Electrode