Learn More
Kyfora Bio PEI branched MW 600 100g
Catalog No. 5000285304
Encompass
Branched Polyethylenimine MW 600 (bPEI 600) is branched liquid water soluble polyamine with a high cationic charge density that readily binds anionic substrates It can be used in the formulation of nanocarriers such as PEI pcoated graphene oxide nanoparticles or PEI coated mesoporous silica nanocarriers for a combination of nucleic acid and drug delivery
Catalog No. 50-002-85304 Supplier Kyfora Bio Supplier No. 02371100
Branched Polyethylenimine MW 600 (bPEI 600) is branched liquid water soluble polyamine with a high cationic charge density that readily binds anionic substrates It can be used in the formulation of nanocarriers such as PEI pcoated graphene oxide nanoparticles or PEI coated mesoporous silica nanocarriers for a combination of nucleic acid and drug delivery