Learn More
Kyfora Bio Polyethylenimine Hydrochloride, Linear MW 4,000 (LPEI 4k) 2GR
Supplier:  Kyfora Bio 248852
Encompass_Preferred
Polyethylenimine HCl, Linear, Mw 4,000 (L-PEI HCl 4000) is a fully hydrolyzed (deacylated), highly water soluble hydrochloride salt form of our Polyethylenimine, Linear, Mw 2,500. Low molecular weight linear PEIs, like L-PEI HCL 4000 can be used for delivery of nucleic acids in vitro. Key application(s): Nucleic acid delivery
Catalog No. 50-255-9820