Learn More
TARGETMOL CHEMICALS INC PEIMINE 25MG
SDP
Catalog No. 502576393
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Peimine (Wanpeinine A) has good anti-inflammatory effects in vivo. 2. Peimine is an inhibitor to inhibit LPS-induced production of inflammatory cytokines by blocking the MAPKs and NF-(kappa)B signaling pathway. purity: 99%
Catalog No. 50-257-6393 Supplier TARGETMOL CHEMICALS INC Supplier No. T6S010725MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Peimine (Wanpeinine A) has good anti-inflammatory effects in vivo. 2. Peimine is an inhibitor to inhibit LPS-induced production of inflammatory cytokines by blocking the MAPKs and NF-(kappa)B signaling pathway. purity: 99%