Learn More
Cayman Chemical PeImIsInhydrochlorIde 50mg
SDP
Supplier:  Cayman Chemical 2643850 MG
Encompass_Preferred
A steroidal alkaloid; inhibits rat lung AChE by 20.2% at 200 μM
Catalog No. 50-293-2888