Learn More
Eurofins Discoverx PEK, active, 10 µg. (14-916)
Supplier:  Eurofins Discoverx 14916
Encompass
N-terminal 6xHis-tagged recombinant, human PEK amino acids 536-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9407