Learn More
Selleck Chemical LLC Pelabresib CPI-0610-5mg
SDP
Catalog No. 502389938
Encompass_Preferred
Pelabresib (CPI-0610) is a potent and selective benzoisoxazoloazepine BET bromodomain inhibitor with an IC50 of 39 nM for BRD4-BD1 in TR-FRET assay and currently undergoing human clinical trials for hematological malignancies. CPI-0610 inhibits the expression of Nuclear receptor binding SET domain protein 3 (NSD3) target genes.
Catalog No. 50-238-9938 Supplier Selleck Chemical LLC Supplier No. S78535MG
Pelabresib (CPI-0610) is a potent and selective benzoisoxazoloazepine BET bromodomain inhibitor with an IC50 of 39 nM for BRD4-BD1 in TR-FRET assay and currently undergoing human clinical trials for hematological malignancies. CPI-0610 inhibits the expression of Nuclear receptor binding SET domain protein 3 (NSD3) target genes.