Learn More
Medchemexpress LLC Pelargonidin-3-O-glucoside chloride | 18466-51-8 | MFCD00017578 | 98.0% | 468.84 g·mol⁻¹ | C21H21ClO10 | 5 MG
SDP
Catalog No. 5000301976
Encompass_Preferred
Callistephin chloride (pelargonidin-3-O-glucoside chloride; CAS 18466-51-8) is an anthocyanin research reagent with antioxidant and signaling-modulating activities. It is employed in biochemical and cell biology studies to investigate oxidative stress, inflammatory pathways, and neuroprotective or retinal-protective effects. Supplied as a chloride salt for in vitro research, handle and store protected from light and moisture.
  • Antioxidant activity and free radical scavenging
  • Modulates inflammatory and apoptosis-related signaling
  • Demonstrated neuroprotective and retinal protective effects in vitro
  • Suitable for biochemical and cell biology research applications
  • Provided as a stable chloride salt for laboratory use
  • Store at 4°C; protect from light and moisture
Catalog No. 50-003-01976 Supplier Medchemexpress LLC Supplier No. HYW0400455MG
Add to Cart
Add to Cart
Callistephin chloride (pelargonidin-3-O-glucoside chloride; CAS 18466-51-8) is an anthocyanin research reagent with antioxidant and signaling-modulating activities. It is employed in biochemical and cell biology studies to investigate oxidative stress, inflammatory pathways, and neuroprotective or retinal-protective effects. Supplied as a chloride salt for in vitro research, handle and store protected from light and moisture.
  • Antioxidant activity and free radical scavenging
  • Modulates inflammatory and apoptosis-related signaling
  • Demonstrated neuroprotective and retinal protective effects in vitro
  • Suitable for biochemical and cell biology research applications
  • Provided as a stable chloride salt for laboratory use
  • Store at 4°C; protect from light and moisture